Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Power Blotter Select Transfer Stacks, nitrocellulose, mini

Catalog No. p-7180367
Encompass_Preferred
Change view
Click to view available options
Sufficient For:
10 Blots
40 Blots
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
PB3210 10 Blots
PB3240 40 Blots
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PB3210 Supplier Invitrogen™ Supplier No. PB3210
Only null left
Add to Cart
Edge
Add to Cart

Transfer proteins rapidly and efficiently with these ready-to-use high performance transfer stacks for Invitrogen Power Blotter.

The Invitrogen Power Blotter Select Transfer Stacks are used to transfer proteins using the Invitrogen Power Blotter System. Disposable, pre-assembled stacks come with an integrated nitrocellulose transfer membrane to perform dry blotting of proteins from polyacrylamide gels. Each Power Blotter Select Transfer Stack contains filter paper, membrane and the appropriate cathode and anode buffers incorporated into a solid gel matrix to allow fast, reliable transfer of proteins.

Nitrocellulose Membrane:

  • 0.2 μm pore size
  • Composed of 100% pure nitrocellulose for high-quality transfer.
  • Compatible with commonly used detection methods such as staining, chemiluminescence, or fluorescence.
  • Proteins bind as hydrophobic and electrostatic interactions.
  • Protein binding capacity is 209 μg/cm2.

Rapid, efficient and even transfer:

  • Ready-to-use transfer stack takes little time to set up
  • Protein transfers in as little as 7 minutes
  • Stacks made with high quality membranes for high binding efficiency, uniform transfer, and high detection sensitivity

Increased reproducibility:

  • Preassembled stacks provide consistent performance

 

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 8 cm
Width (Metric) 8 cm
Quantity 10 stacks
Format Transfer Stack
Material Nitrocellulose
Pore Size 0.2 μm
Content And Storage 10 mini-size transfer stacks (8 cm x 8 cm)
Shipping Condition Room Temperature
Dimensions (LxW) Mini, 8 cm x 8 cm
Product Line Power Blotter
Sufficient For 10 Blots
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.