Learn More
Invitrogen™ Power Blotter Select Transfer Stacks, nitrocellulose, regular size
Description
The Invitrogen Power Blotter Select Transfer Stacks are used to transfer proteins using the Invitrogen Power Blotter System. Disposable, pre-assembled stacks come with an integrated nitrocellulose transfer membrane to perform dry blotting of proteins from polyacrylamide gels. Each Power Blotter Select Transfer Stack contains filter paper, membrane and the appropriate cathode and anode buffers incorporated into a solid gel matrix to allow fast, reliable transfer of proteins.
Nitrocellulose Membrane:
- 0.2 μm pore size
- Composed of 100% pure nitrocellulose for high-quality transfer.
- Compatible with commonly used detection methods such as staining, chemiluminescence, or fluorescence.
- Proteins bind as hydrophobic and electrostatic interactions.
- Protein binding capacity is 209 μg/cm2.
Rapid, efficient and even transfer:
- Ready-to-use transfer stack takes little time to set up
- Protein transfers in as little as 7 minutes
- Stacks made with high quality membranes for high binding efficiency, uniform transfer, and high detection sensitivity
Increased reproducibility:
- Preassembled stacks provide consistent performance
Specifications
Specifications
| Length (Metric) | 13 cm |
| Width (Metric) | 8.3 cm |
| Quantity | 10 stacks |
| Format | Transfer Stack |
| Material | Nitrocellulose |
| Pore Size | 0.2 μm |
| Content And Storage | 10 regular-size transfer stacks (13 cm x 8.3 cm) |
| Shipping Condition | Room Temperature |
| Dimensions (LxW) | Regular, 13 cm x 8.3 cm |
| Product Line | Power Blotter |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.