Learn More
Grainger Power Correction Capacitor 20 kvar
Supplier:  Grainger 16Y199
Encompass_Preferred
The WEG Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a kVAR rating of 25 kVAR with 480V voltage. It features a heavy duty canister construction and is UL listed. The dimensions are 18" diameter x 39" height. This capacitor helps reduce energy costs and prevent overheating and inefficiency in motors, transformers, and electrical systems by correcting power factor issues. It is well-suited for industrial and commercial applications.
Catalog No. 50-276-6212
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.