Learn More
Grainger Power Correction Capacitor 30 kvar
Supplier:  Grainger 16Y138
Encompass_Preferred
The Grainger 16Y138 is a Westward 3/8-inch drive deep impact socket set. This 12-piece socket set includes sizes ranging from 10mm to 19mm, providing a versatile range of metric sockets for impact wrench use. The sockets are made from chrome vanadium steel for durability and feature a black phosphate finish to resist corrosion. The deep design allows access to fasteners in tight spaces. Overall this Westward impact socket set offers a quality selection of 3/8-inch drive metric sockets for impact tools.
Catalog No. 50-276-5219
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.