Learn More
Power Hawk DC-1 Damage Control Kit
Catalog No. 19817192 Shop All Power Hawk Technologies, Inc. Products
Click to view available options
Product Type:
Damage Control Kit
Catalog No. Product Type
19-817-192 Damage Control Kit
1 options
Catalog No. 19-817-192 Supplier Power Hawk Supplier No. DC1
Add to Cart
Add to Cart
Request a Quote
Includes
  • P-16 Rescue Tool (1)
  • AP-1600 Attachment pins (1)
  • CA-4M Power Cable (1)
  • PC-100 Controller Unit (1)
  • PWR-3X12 Power Packs (2)
  • JC4-16 Jumper Cables (1)
  • BC-U1 Battery Charger (1)
  • S-1601 Spreader Arms (1)
  • C-1601 Curved-Blade Cutter (1)
  • CS-1602 Straight-Blade Cutter (1)
  • P-25 Power Push Ram (1)
  • P-40 Power Push Ram (1)
  • E-24 in. Ram Extension (1)
  • E-48 in. Ram Extension (1)
  • 2C0516-1 Ram Couplings (2)
  • 2C0551-1 Ram Swivel Set (1)
  • LB-1204 Floodlight
  • EC4-16 Extension Cable, 16 ft. (1)
  • BKPK-100 Back Pack (1)
  • SZA-1015 Sawzall™ Extrication Kit (1 each - kit includes 18VDC cordless Sawzall, SZA1812-15 power adapter, EC4-16 extension cable, five torch blades, one ax blade, and carrying bag)
All-electric, jaws-type system operates completely without hydraulics for exceptional versatility and power
Developed for military fighter jet control applications using aerospace technolog. Highly effective for use in numerous rescue applications including, vehicle extrication, collapse and remote rescues, indoor and confined-space operations, forcible entry and special operations.
  • Lightweight and compact for portability and ease of use
  • No set-up necessary for rapid response
  • Safe for indoor/outdoor use—runs in O2 depleted atmosphere
  • Interchangeable spreaders and cutters maximize versatility and minimize compartment space
  • Simultaneously operates accessories such as Milwaukee™ Sawzall™, Ramfan™ blowers, lights and more with 12VDC power pack
  • Ambidextrous controls; color-coded for enhanced safety and ease of operation
  • Patented 70° swivel-head for around-the-corner purchase points
  • Operates quietly and requires little maintenance
PEARS (Portable Electric All-Purpose Rescue Set)
  • Assessed and approved through the U.S. Navy's Non-Developmental Items Program (NDI)
  • Replaces PHARS (Portable Hydraulic Access and Rescue Set) on U.S. Navy ships
  • Outfitted on new construction ships, PEARS is the only NAVSEA authorized replacement for PHARS on existing ships
Power Hawk Rescue System components and accessories available in various kit configurations and individually.
Certifications
UL Certified Compliant to NFPA 1936 (P-16 Rescue Tool w/S-1601, C-1601, CS-1602, P-25, and P-40 Rams, PWR-3X12 Power Pack).
Specifications
Product Type Damage Control Kit
Includes P-16 rescue tool, AP-1600 attachment pins, CA-4M power cable, PC-100 controller unit, PWR-3X12 power pack, JC4-16 jumper cables, BC-U1 battery charger, S-1601 spreader arms, C-1601 curved-blade cutter, CS-1602 straight-blade cutter, P-25 power push ram, P-40 power push ram, E-24 in. ram extension, E-48 in. ram extension, 2C0516-1 ram couplings, 2C0551-1 ram swivel set, LB-1204 floodlight, EC4-16 extension cable, 16 ft., BKPK-100 back pack