Learn More
Power Hawk P-16 Raptor Kit
Catalog No. 19817203 Shop All Power Hawk Technologies, Inc. Products
Click to view available options
Product Type:
Raptor Kit
Catalog No. Product Type
19-817-203 Raptor Kit
1 options
Catalog No. 19-817-203 Supplier Power Hawk Supplier No. RAPTOR
Add to Cart
Add to Cart
Request a Quote
Includes
Rescue tool, attachment pin set, power cable, controller, two power packs (12V), jumper cables, battery charger, spreader arm set, curved- abd straight-blade cutters, 25 and 40 in. rams and coupler, floodlight, extension cable (16 ft.).
The versatility of the Power Hawk makes it the perfect choice for a wide range of rescue operations
Extremely powerful and uniquely versatile rescue tool eliminates the use of hydraulics completely and utilizes 12 Volts DC power. This all-electric jaws-type rescue tool utilizes Curtiss-Wright aerospace gear technology developed for demanding military fighterjet flight controls.
  • No hydraulics
  • Lightweight and portable
  • Rapid response; no setup
  • Runs in O2 depleted atmosphere
  • Quiet operation
  • Low maintenance
  • Compact, saves space
  • Interchangeable spreaders and cutters
  • 70-degree pivot of powerhead
  • Power-blade cross-over feature
  • 12-volt back-up readily available
  • Simultaneously run accessories such as the Milwaukee Sawzall™, RamFan blower, lights, winches and more
Specifications
Product Type Raptor Kit
Includes P-16 rescue tool, AP-1600 attachment pins, CA-4M power cable, PC-100 controller unit, PWR-3X12 power pack, JC4-16 jumper cables, BC-U1 battery charger, S-1601 spreader arms, C-1601 curved-blade cutter, CS-1602 straight-blade cutter, P-25 power push ram, P-40 power push ram, E-24 in. ram extension, E-48 in. ram extension, 2C0516-1 ram couplings, 2C0551-1 ram swivel set, LB-1204 floodlight, EC4-16 extension cable, 16 ft.