Learn More
Power Hawk SH-1 Super Hornet Kit
Catalog No. 191305320 Shop All Power Hawk Technologies, Inc. Products
Click to view available options
Product Type:
Super Hornet Kit
Catalog No. Product Type
19-130-5320 Super Hornet Kit
1 options
Catalog No. 19-130-5320 Supplier Power Hawk Supplier No. SH1
Add to Cart
Add to Cart
Request a Quote
Includes
  • P-16 Rescue Tool (1)
  • AP-1600 Attachment pins (1)
  • CA-4M Power Cable (1)
  • PC-100 Controller Unit (1)
  • PWR-3X12 Power Packs (2)
  • JC4-16 Jumper Cables (1)
  • BC-U1 Battery Charger (1)
  • S-1601 Spreader Arms (1)
  • C-1601 Curved-Blade Cutter (1)
  • CS-1602 Straight Blade Cutter (1)
  • SZA-1015 Sawzall™ Extrication Kit (1 each - kit includes 18 VDC cordless Sawzall, SZA1812-15 power adapter, EC4-16 extension cable, five torch blades, one ax blade, and carrying bag)
Provides rapid response cutting and spreading capabilities for access into aircraft
Designed with the needs of U.S. Navy Flight Deck crews in mind. Assessed and approved through the U.S. Navy Non-Developmental Items Program (NDI). Utilizing aerospace technology developed for military fighter jet control applications, this all-electric, jaws-type system operates completely without hydraulics for exceptional versatility and power. Highly effective for use in numerous rescue applications including, vehicle extrication, collapse and remote rescues, indoor and confined-space operations, forcible entry and special operations.
  • Lightweight and compact for portability and ease of use
  • No set-up necessary for rapid response
  • Safe for indoor/outdoor use—runs in O2 depleted atmosphere
  • Interchangeable spreaders and cutters maximize versatility and minimize compartment space
  • Simultaneously operates accessories such as Milwaukee™ Sawzall™, Ramfan™ blowers, lights and more with 12VDC power pack
  • Ambidextrous controls; color-coded for enhanced safety and ease of operation
  • Patented 70° swivel-head for around-the-corner purchase points
  • Operates quietly and requires little maintenance
Power Hawk Rescue System components and accessories available in various kit configurations and individually.
Certifications
UL Certified Compliant to NFPA 1936 (P-16 Rescue Tool w/S-1601, C-1601, CS-1602, P-25, and P-40 Rams, PWR-3X12 Power Pack).
Specifications
Product Type Super Hornet Kit
Includes P-16 rescue tool, AP-1600 attachment pins, CA-4M power cable, PC-100 controller unit, PWR-3X12 power pack, BC-U1 battery charger, S-1601 spreader arm, C-1601 curved-blade cutter, CS-1602 straight blade cutter, SZA-1015 Sawzall™ extricati