Learn More
Cole-Parmer Extech 382202 Single Output Economical DC Power Supply, 18V/3A
Supplier:  Cole-Parmer 2684980
Encompass_Preferred
These economical DC power supplies are ideal for bench testing, field service, R & D, educational, and telecommunication applications. Units provide short circuit and overload protection and display both voltage and current on the dual backlit display. Additional features include adjustable constant voltage or constant current output, positive and negative banana plug output jacks, and an illuminated on/off switch.
Catalog No. 50-993-010
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.