Learn More
Sartorius Power Supply with Plug-In AC Adapter Set
Catalog No. 14558929 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Balance's Power Supply
Catalog No. For Use With (Equipment)
14-558-929 Balance's Power Supply
1 options
This item is not returnable. View return policy
Catalog No. 14-558-929 Supplier Sartorius Supplier No. YEPS01PS4
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sartorius power supply with country-specific plug-in AC adapters.
  • Sartorius power supply with country-specific plug-in AC adapters
    • Primary: 100 to 240 V, 50/60 Hz, 0.2 A
    • Secondary: 15 V DC, 0.53 A (max.)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Balance's Power Supply
Type Power Adapter