Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PowerEase™ Touch 120W Power Supply and XCell SureLock™ Mini-Cell and XCell II™ Blot Module Welcome Pack

Catalog No. PSC120SB
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PSC120SB 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PSC120SB Supplier Invitrogen™ Supplier No. PSC120SB
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The PowerEase Touch 120W Power Supply and XCell SureLock Mini-Cell and XCell II Blot Module Welcome Pack contains: PowerEase Touch 120W Power Supply and XCell SureLock Mini-Cell and XCell II Blot Module

The PowerEase Touch 120W Power Supply and XCell SureLock Mini-Cell and XCell II Blot Module Welcome Pack contains:

  • PowerEase Touch 120W Power Supply
  • XCell SureLock Mini-Cell and XCell II Blot Module

About the PowerEase Touch 120W Power Supply

The Invitrogen PowerEase Touch 120W Power Supply is a medium-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for DNA/RNA electrophoresis, SDS-PAGE, and native PAGE. The PowerEase Touch 120W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.

About the XCell SureLock Mini-Cell and XCell II Blot Module

The XCell SureLock Mini-Cell and XCell II Blot Module set is an electrophoresis and blotting system for mini gels. It includes the XCell SureLock Mini-Cell for running Invitrogen mini gels and the XCell II Blot Module for blotting up to two mini gels at a time. The XCell SureLock Mini-Cell holds gels firmly with a simple gel tension wedge. When the lever on the gel tension wedge is pushed forward into the locked position, an even, horizontal force is generated. This seals the gel/buffer core assembly firmly into position in the lower buffer chamber. The positive locking action of the gel tension wedge ensures a trouble-free, leak-free gel run every time.

The XCell II Blot Module allows you to easily transfer proteins or nucleic acids from mini gels to membranes. It fits into the XCell SureLock and XCell II Mini-Cells in place of the gel/buffer core assembly. It requires less than 200 mL of transfer buffer for western, southern, and northern transfers. Tough platinized titanium and stainless steel electrodes create a uniform electrical field without clamps or hinged gel holders. Maximum blot size is 9 cm x 9 cm.

TRUSTED_SUSTAINABILITY

Specifications

Ambient Temperature (English) Room Temperature
For Use With (Application) DNA/RNA electrophoresis, SDS-PAGE, and native PAGE
Gel Size Mini
Max. Current 500 mA
Product Line PowerEase
Timer Up to 999 min./step
Display 4.3 inch backlit LCD
No. of Jacks 4 (4 positive voltage and 4 negative voltage)
Quantity 1 pack
Shipping Condition Room temperature
Stackable Yes
Warranty One year
Dimensions (L x W x H) 30.3 inch x 23 inch x 10.2 inch
Weight 2.1 kg
Max. Voltage 300 VDC
Wattage 120 watts
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.