Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PowerEase™ Touch 350W Power Supply and SureLock™ Tandem Midi Gel Tank Welcome Pack

Catalog No. PSC350TT
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PSC350TT 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PSC350TT Supplier Invitrogen™ Supplier No. PSC350TT
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The PowerEase Touch 350W Power Supply and SureLock Tandem Midi Gel Tank Welcome Pack contains: PowerEase Touch 350W Power Supply and SureLock Tandem Midi Gel Tank

The PowerEase Touch 350W Power Supply and SureLock Tandem Midi Gel Tank Welcome Pack contains:

  • PowerEase Touch 350W Power Supply
  • SureLock Tandem Midi Gel Tank

About the PowerEase Touch 350W Power Supply

The Invitrogen PowerEase Touch 350W Power Supply is a high-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for high current applications like high throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, and western blotting. The PowerEase Touch 350W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.

About the SureLock Tandem Midi Gel Tank

The Invitrogen SureLock Tandem Midi Gel Tank provides for rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a setup time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.

TRUSTED_SUSTAINABILITY

Specifications

Ambient Temperature (English) Room Temperature
For Use With (Application) High throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE and western blotting
Gel Size Midi
Max. Current 3 A
Product Line PowerEase
Timer Up to 999 min./step
Display 4.3 inch backlit LCD
No. of Jacks 4 (4 positive voltage and 4 negative voltage)
Quantity 1 pack
Shipping Condition Room Temperature
Stackable Yes
Warranty One year
Dimensions (L x W x H) 30.3 inch x 23 inch x 10.2 inch
Weight 2.1 kg
Max. Voltage 300 VDC
Wattage 350 watts
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.