Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ PowerFlex™ Thermal Cycler, 3x32-well with Total 3 Years of Depot Repair Center Service Coverage and 1 Onsite Multiplex Dynamic Temperature Verification (MDTV) Preventive Maintenance (PM)

Catalog No. 5000575227
Encompass_Preferred
Change view
Click to view available options
3 x 32-well, 2-Zone VeriFlex Blocks, Total 3 Years of Depot Repair Center Service Coverage, 1 Onsite Multiplex Dynamic Temperature Verification (MDTV) Preventive Maintenance (PM)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-005-75227 3 x 32-well, 2-Zone VeriFlex Blocks, Total 3 Years of Depot Repair Center Service Coverage, 1 Onsite Multiplex Dynamic Temperature Verification (MDTV) Preventive Maintenance (PM)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-005-75227 Supplier Applied Biosystems™ Supplier No. POWERFLEX32RCMDTV
Only null left
Add to Cart
Add to Cart

The Applied Biosystems™ PowerFlex™ Thermal Cycler is a high-performance PCR thermal cycler designed for precise temperature control, reproducible amplification, and flexible throughput in research laboratories.

The Applied Biosystems™ PowerFlex™ Thermal Cycler is a high-performance PCR thermal cycler designed for precise temperature control, reproducible amplification, and flexible throughput in research laboratories. The PowerFlex™ Thermal Cycler builds on the widely adopted ProFlex™ PCR System platform and incorporates enhanced thermal accuracy, faster ramp rates, multiple block configurations, and modern connectivity features to support high-throughput and multi-user PCR workflows.

The PowerFlex™ Thermal Cycler is available in 96-well and 3 x 32-well formats. The system is suitable for a wide range of PCR applications, including cloning, genotyping, sequencing, mutagenesis, automation workflows, and high-throughput screening.

PowerFlex™ Thermal Cycler features

  • Temperature accuracy of ±0.15°C (35–99.9°C)
  • Well-to-well uniformity of <0.4°C at 95°C and <0.3°C at 72°C
  • Maximum ramp rate of 7.0°C/sec
  • VeriFlex™ Blocks technology for precise PCR optimization
  • Independent block control (3 x 32-well format)
  • Simulation Mode for protocol transfer
  • 10.1-inch touchscreen user interface
  • Smart Help integrated service support
  • Compatibility with fully skirted, semi-skirted, and non-skirted PCR plates

Precise thermal control for reproducible PCR

The PowerFlex™ Thermal Cycler is engineered to meet stringent PCR temperature control requirements. With ±0.15°C temperature accuracy and tight well-to-well uniformity, the system supports consistent amplification across the entire block. Uniform heating and cooling help reduce variability between wells, which is critical for comparative analyses and high-throughput workflows.

The maximum ramp rate of 7.0°C/sec reduces overall PCR run time while maintaining amplification performance and data integrity.

VeriFlex™ Blocks technology for PCR optimization

The PowerFlex™ Thermal Cycler incorporates Applied Biosystems™ VeriFlex™ Blocks technology. The 96-well configuration includes up to six independently controlled temperature zones across a 30°C range. Users can define minimum and maximum temperatures, select a midpoint, or program individual zone temperatures.

Unlike traditional gradient blocks that estimate intermediate temperatures, VeriFlex™ Blocks allow direct control of defined temperature zones, enabling precise and reproducible PCR optimization.

The 3 x 32-well configuration includes independently controlled blocks, allowing multiple protocols to run simultaneously.

Flexible block configurations

The PowerFlex™ Thermal Cycler supports laboratories with varying throughput requirements:

  • 96-well, 0.2 mL block format
  • 3 x 32-well, 0.2 mL block format with independent control
  • 2 x 384-well format with independent control (available later)

The 3 x 32-well format enables 3 users or protocols to be run within the same instrument. This configuration is particularly suited for shared laboratories, core facilities, and workflows requiring concurrent assay development or multiple users.

Simulation Mode for protocol transfer

Simulation Mode allows the PowerFlex™ Thermal Cycler to emulate ramp rates and cycling behavior from legacy Applied Biosystems thermal cyclers and selected competitor systems. This feature helps reduce or eliminate the need for protocol re-optimization when transitioning to a new instrument.

Simulation Mode supports workflow continuity and simplifies instrument upgrades within established laboratories.

User interface and connectivity

The PowerFlex™ Thermal Cycler features a 10.1-inch thin-film transistor (TFT) touchscreen with an intuitive graphical user interface for protocol setup, editing, and monitoring.

The system includes:

  • Onboard protocol storage
  • USB connectivity
  • Ethernet connectivity
  • Optional Wi-Fi connectivity using the Applied Biosystems™ High-Power USB Wi-Fi Module (Cat. No. A26774)

Cloud-enabled connectivity through the Thermo Fisher™ Connect Platform allows remote protocol management, scheduling, and run monitoring.

Smart Help and service support

Smart Help functionality enables direct submission of technical support or service requests from the PowerFlex™ Thermal Cycler. Instrument log files and run history can be attached automatically to facilitate troubleshooting and accelerate resolution.

Service plans are available to support long-term instrument performance, including:

  • AB Repair Center Service
  • Rapid Exchange Service
  • Multiplex Dynamic Temperature Verification (MDTV) and Planned Maintenance
  • Qualification services (IQ/OQ)

Reliability and sustainability

PowerFlex™ Thermal Cyclers are manufactured using components that undergo environmental and mechanical stress testing to support long-term reliability.

The instrument design incorporates sustainability considerations, including reduced energy consumption through sleep mode functionality, use of recycled materials in selected components and recyclable components in the packaging.

TRUSTED_SUSTAINABILITY

Specifications

Block Format Fixed
Thermal Accuracy ±0.15°C
Reaction Volume Range 1 to 100 μL
Content And Storage 1 Instrument with Service Packages
Thermal Uniformity <0.4°C
Capacity 3 x 32-well
Voltage 100/240 V ±10%
Peak Block Ramp Rate 7°C/sec.
Display Type 10.1 in. LCD Touchscreen
Product Type Thermal Cycler
Height (English) 10.1 in.
Depth (English) 20.2 in.
Height (Metric) 25.7 cm
Warranty 2 Years
Depth (Metric) 51.3 cm
Thermal Range 0°C to 100°C
For Use With (Application) End Point PCR
Length (English) 11.8 in.
Length (Metric) 30 cm
No. of Programs 1000
Heated Lid Yes
Quantity 1 Ea.
Total Service Coverage Total 3 Years of Depot Repair Center Service Coverage and 1 Onsite Multiplex Dynamic Temperature Verification (MDTV) Preventive Maintenance (PM)
Block Configurations 3 x 32-well, 2-Zone VeriFlex Blocks
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.