Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech PowerStem MSC1, Serum-free culture medium kit for human mesenchymal stem cells (hMSC), 20 x 500 ml kit
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0477355KX20
Encompass_Preferred
PAN-Biotech PowerStem MSC1: Serum-free culture medium kit for human mesenchymal stem cells (hMSC). QUANTITY: 20 kits (500 ml/each). MANUFACTURED BY: Pan-Biotech (Germany). DESCRIPTION: PAN-Biotech PowerStem MSC1 is an easy to use xeno-free, serum-free medium for in vitro proliferation of human mesenchymal stem cells (hMSC). APPLICATION: Serum-free cultivation of human mesenchymal stem cells (hMSC) while maintaining the undifferentiated state and multi-lineage potential. By adding specific differentiation factors, MSC can be differentiated in vitro to the desired cell types (bone, cartilage, adipose tissue etc.). COMPOSITION: PowerStem MSC1 contains salts, amino acids, trace elements, hormones, growth factors, enriched human proteins and lipids in an optimized formulation. KIT CONTENTS: PowerStem MSC1 basal medium (470 ml, Cat. No. P04-77350C), PowerStem MSC1 growth supplement 1 (5 ml, Cat. No. P04-77350S1), PowerStem MSC1 growth supplement 2 (25 ml, Cat. No. P04-77350S2).
Catalog No. 50-238-8640
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.