Learn More
Cytiva (Formerly Pall Lab) Polypropylene/Polyethylene Separator Depth Filters - 10 m, 25 mm (500/pkg)
Catalog No. 502063248 Shop All Cytiva Products
Encompass
Virtually Inert to Chemical and Biological Agents-Outstanding wet strength-High chemical resistance-Provides a channeling effect for liquid, increasing overall filter flow rate when used in series with submicron membrane filters-Ensures stability when filtering concentrated acids, alkalis, or oxidizers This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3248 Supplier Cytiva Supplier No. 60343
Virtually Inert to Chemical and Biological Agents-Outstanding wet strength-High chemical resistance-Provides a channeling effect for liquid, increasing overall filter flow rate when used in series with submicron membrane filters-Ensures stability when filtering concentrated acids, alkalis, or oxidizers This product is not approved for use in medical, clinical, surgical or other patient applications.