Learn More
United Scientific Supplies PP/PTFE Stopcocks, 6 Pk.
SDPSCI_ED
Catalog No. S39222 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Description:
Three-way
Two-way
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
S39222 Three-way
S39221 Two-way
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S39222 Supplier United Scientific Supplies Supplier No. 55302
Add to Cart
Add to Cart
United Scientific PP/PTFE Stopcocks are leak-proof and available in both two and three way
  • Leak-proof, economical polypropylene stopcocks with PTFE plug
  • Featuring serrated ports that accept tubing from 6 mm to 10 mm in diameter
Specifications
Type Stopcock
Plug Bore 6 to 10 mm
Description Three-way
Material Polypropylene, PTFE
Autoclavable Autoclavable
For Use With (Application) Stopcock for Lab Tubing
For Use With (Equipment) Lab Tubing
Includes Stopcock
Plug Material PTFE
Outlet Port Serrated
Connectors Serrated Ports