Learn More
Anton Paar 12 ml Polypropylene vials (2000 pcs)
Supplier: Anton Paar 21550
Pack of 2000 plastic vials used with a sample changer (Anton Paar Xsample series) to automatically fill up to 12 mL of sample into your Anton Paar measuring instrument.Recommended Cap Standard Cap for 12 ml Vials NC9145272
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.