Learn More
PP2A Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
Specifications
Specifications
| Antigen | PP2A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | PPP2CB |
| Gene Accession No. | P62714, P62715 |
| Gene Alias | PP2Abeta, PP2CB, PP2A-beta, protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform, protein phosphatase 2, catalytic subunit, beta isoform, protein phosphatase 2A catalytic subunit, beta isoform, protein phosphatase type 2A catalytic subunit, serine/threonine pro |
| Gene Symbols | PPP2CB |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.