Learn More
Description
PP2CA/PP2CB Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PP2CA/PP2CB |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | Ppm1b |
| Gene Accession No. | O62829, O62830, O75688, P20650, P35813, P35815, P36993, P49443 |
| Gene Alias | 2310003C21Rik; 2900017D14Rik; AI427932; AU017636; FLJ42306; IA; MGC9201; MMPa-2; Mpp alpha; MPPa-1; Pp2c1; Pp2c2; PP2CA; PP2Calpha; PP2C-ALPHA; PP2CB; PP2CBETA; PP2C-beta; PP2C-beta-X; PPC2BETAX; Ppm1a; PPM1B; PPM1B protein; Pppm1a; Pppm1b; Protein phosphatase 1A; protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform; protein phosphatase 1A, magnesium dependent, alpha isoform; protein phosphatase 1B; protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform; protein phosphatase 1B, magnesium dependent, beta isoform; protein phosphatase 2C alpha; protein Phosphatase 2C beta; protein phosphatase 2C isoform alpha; Protein phosphatase 2C isoform beta; protein phosphatase 2C-like protein; protein phosphatase IA; Protein phosphatase type 1A (formely 2C) Mg-dependent alpha isoform; Protein phosphatase type 1A (formely 2C), Mg-dependent, alpha isoform; Protein phosphatase type 1B (formely 2C) Mg-dependent beta isoform; Protein phosphatase type 1B (formely 2C), Mg-dependent, beta isoform; protein phosphatase, Mg2+/Mn2+ dependent 1A; protein phosphatase, Mg2+/Mn2+ dependent 1B; protein phosphatase, Mg2+/Mn2+ dependent, 1A; protein phosphatase, Mg2+/Mn2+ dependent, 1B; Ser/Thr protein phosphatase type 2C beta 2 isoform |
| Gene Symbols | PPM1A, Ppm1b |
| Host Species | Sheep |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
