Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPAP2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA112730
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA112730 100 μg
1 options

Catalog No. PIPA112730

Supplier: Invitrogen™ PA112730

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for this product is human brain lysate.

Human vascular endothelial growth factor produced in E.Coli is a double, non-glycosylated, polypeptide chain containing 165 amino acids and having a molecular mass of 38231 Dalton. The rHuVEGF is purified by proprietary chromatographic techniques.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAP2B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene PLPP3
Gene Accession No. O14495, P97544, Q99JY8
Gene Alias 1110003O22Rik; 2610002D05Rik; AV025606; D4Bwg0538e; D4Bwg1535e; differentially expressed in rat intestine 42; Dri42; EC 3.1.3.4; ER transmembrane protein Dri 42; lipid phosphate phosphohydrolase 3; Lpp3; MGC15306; PAP2 beta; PAP2B; PAP-2b; PAP2-b; PAP2-beta; phosphatidate phosphohydrolase type 2b; Phosphatidic acid phosphatase 2b; phosphatidic acid phosphatase type 2B; phospholipid phosphatase 3; PLPP3; Ppab2b; PPAP2B; type-2 phosphatidic acid phosphatase-beta; vascular endothelial growth factor and type I collagen inducible; Vascular endothelial growth factor and type I collagen-inducible protein; VCIP
Gene Symbols PLPP3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to phosphatidic acid phosphatase 2B.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192270, 67916, 8613
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.