Learn More
Description
Keep as concentrated solution. Predicted reactivity: Mouse (91%), Rat (88%), Dog (92%), Pig (92%), Rhesus Monkey (98%), Bovine (88%), Guinea pig (87%). Positive Control: Rat primary hepatocyte, mouse liver, mouse brain, mouse lung. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PPAR alpha |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.44 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | PPARA |
| Gene Accession No. | P23204, P37230, Q07869 |
| Gene Alias | 4933429D07Rik; AW742785; hPPAR; I79_022392; MGC2237; MGC2452; Nr1c1; nuclear hormone receptor; nuclear receptor; nuclear receptor subfamily 1 group C member 1; peroxisome proliferative activated receptor alpha; peroxisome proliferative activated receptor, alpha; peroxisome proliferative activated receptor, alpha isoform 1; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor alpha; peroxisome proliferator-activated nuclear receptor alpha variant 3; peroxisome proliferator-activated receptor alpha; PPAR; PPAR ALPHA; Ppara; PPARALPHA; PPAR-alpha; proliferator-activated receptor alpha; transcription factor belonging to the nuclear receptor superfamily |
| Gene Symbols | PPARA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
