Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPAR gamma-2 Polyclonal Antibody, Alexa Fluor™ 647
GREENER_CHOICE

Catalog No. PIPA1824A64 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1824A64 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA1824A64 Supplier Invitrogen™ Supplier No. PA1824A647
  • $498.52
    $406.65 / Each of 1

    Save $91.87

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-824 detects peroxisome proliferator activated receptor (PPAR) gamma 2 from human and mouse tissues. This antibody does not detect PPAR alpha or PPAR delta. PA1-824 immunogen is a synthetic peptide corresponding to residues M(1) G E T L G D S P I D P E S D S(16) C of human PPAR gamma-2. This sequence is not conserved in PPAR gamma-1.

Peroxisome proliferators are non-genotoxic carcinogens which are purported to exert their effect on cells through their interaction with members of the nuclear hormone receptor family, termed peroxisome proliferator activated receptors (PPARs). Nuclear hormone receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate ligand. Studies indicate that PPARs are activated by peroxisome proliferators such as clofibric acid, nafenopin, and WY-14,643, as well as by some fatty acids. It has also been shown that PPARs can induce transcription of acyl coenzyme A oxidase and cytochrome P450 A6 (CYP450 A6) through interaction with specific response elements. PPAR alpha is activated by free fatty acids including linoleic, arachidonic, and oleic acids. Induction of peroxisomes by this mechanism leads to a reduction in blood triglyceride levels. PPAR alpha is expressed mainly in skeletal muscle, heart, liver, and kidney and is thought to regulate many genes involved in the beta-oxidation of fatty acids. Activation of rat liver PPAR alpha has been shown to suppress hepatocyte apoptosis. PPAR gamma 2, like several other nuclear hormone receptors, heterodimerizes with retinoic X receptor (RXR) alpha to form a transcriptionally competent complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAR gamma-2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Alexa Fluor 647
Formulation PBS with 1.0mg/mL BSA and 0.05% sodium azide
Gene PPARG
Gene Accession No. P37231, P37238
Gene Alias CIMT1; GLM1; Nr1c3; nuclear receptor subfamily 1 group C member 3; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor gamma; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator-activated nuclear receptor gamma variant 1; peroxisome proliferator-activated receptor gamma; PPARG; PPARG1; PPARG2; PPARgamma; PPAR-gamma; PPARgamma2; PPAR-gamma2
Gene Symbols PPARG
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) G E T L G D S P I D P E S D S(16) C of human PPAR gamma-2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19016, 5468
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.