Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Ppar Gamma Antibody, Rabbit Multiclonal, BSA and Azide Free, 1 mg

Catalog No. 502711508
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-1508 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-1508 Supplier Abcam Supplier No. AB3169821MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

Carrier-free, conjugation-ready rabbit multiclonal antibody for flow cytometry, immunoprecipitation, and Western blot analysis.

  • Host species: Rabbit
  • Clonality: Multiclonal
  • Isotype: IgG
  • Applications: Flow Cyt (Intra), WB, IP
  • Reactive species: Human, Mouse, Rat
  • Purification technique: Affinity purification Protein A
  • Form: Liquid
  • Storage buffer: pH: 7.2 to 7.4, Constituents: PBS
  • Additional attributes: Carrier free, Conjugation ready

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule PPAR gamma
Label or Dye Unconjugated
Buffer PBS, pH7.2 to 7.4
Detection Location In-Blot Detection, In-Cell Detection
Form Liquid
Cross Reactivity Human, Mouse, Rat
Formulation PBS, pH 7.2 to 7.4
Quantity 1 mg
For Use With (Application) Flow Cytometry, IP, Western Blot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.