Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAT Monoclonal Antibody (OTI3C6), TrueMAB™, OriGene
SDP

Catalog No. 502540312
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0312 30 μL
50-167-5146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0312 Supplier OriGene Supplier No. TA504457S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PPAT Monoclonal antibody specifically detects PPAT in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen PPAT
Applications Western Blot
Classification Monoclonal
Clone OTI3C6
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Psmb7
Gene Accession No. P70195, Q99436
Gene Alias 5730454C12Rik; AA408689; AA675351; AI507113; Amidophosphoribosyltransferase; amidophosphoribosyltransferase proprotein; amidophosphoribosyltransferase-like protein; ATASE; C79945; EC 2.4.2.14; EGK_15868; glutamine phosphoribosylpyrophosphatate amidotransferase; glutamine phosphoribosylpyrophosphate amidotransferase; glutamine PRPP amidotransferase; GPAT; phosphoribosyl pyrophosphate amidotransferase; PPAT; PRAT
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 58-277 of human PSMB7 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19177, 5695
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.