Learn More
HF Scientific™ PPD-2 DPD Powder Pop™ Dispensers for Chlorine Testing
Description
Portable dispenser accurately delivers the correct volume of DPD reagent for Free or Total Chlorine—use with any DPD-based photometer or test kit.
- Supplied with enough DPD reagent for 400 tests
- To use, simply invert dispenser over sample and press button to dispense reagent; powder dissolves quickly in sample, leaving no messy residue behind
- Available for 5 and 10mL sample sizes, with formulations for a variety of photometers and test kits
Specifications
Specifications
| Volume (Metric) | 5 mL |
| For Use With (Application) | Chlorine Testing |
| For Use With (Equipment) | HF Scientific, Hach |
| Includes | Pocket photometer |
| Quantity | 400 Tests |
| Type | Chlorine Reagent Powder Pop Dispenser |
| Volume (English) | 0.16 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.