Learn More
Poltex PPE Station 2 - Gown, Wall Mount
SDP
Catalog No. 22022546
Change view
Click to view available options
PPE Dispenser
Catalog No.
22-022-546 PPE Dispenser
1 options
Catalog No. 22-022-546 Supplier Poltex Supplier No. PPESTAT2W
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
This PPE Isolation Station holds disposable gowns, two boxes of ear loop masks, and gloves. Mounts to wall with keyhole slots. Wall mount (screws/anchors included).
  • PPE Isolation Station holds disposable gowns, two boxes of ear loop masks, and gloves
  • Durable PETG construction
  • Mounts to wall with keyhole slots (screws/anchors included for wall mount)
Specifications
Product Type PPE Dispenser
Color Clear
Length (Metric) 457.2 mm
Width (Metric) 95.25 mm
Height (Metric) 457.20 mm
Holds Bouffant Caps, Ear Plugs, Glasses, Shoe Covers
Material PETG
Mounting Wall Mount
No. of Compartments 2
Product Style Side Loading
Sterility Non-sterile