Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex PPE Station - Gown, Mask, Glove - Wall Mount
SDP

Catalog No. 01259132
Change view
Click to view available options
Dimensions (L x W x H):
4 x 18 x 20 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
01-259-132 4 x 18 x 20 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
N/A
Catalog No. 01-259-132 Supplier Poltex Supplier No. PPESTAT7W
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N/A

PPE isolation station that holds a variety of PPE. Choose from wall mount or over the door hanger options with durable PETG construction.

  • PPE isolation station holds a variety of PPE
  • Available in wall mount (mounting hardware included)
  • Available with over-the-door hanger (hanger included)
  • Durable and cleanable PETG construction
  • Consumables not included

Specifications

Product Type PPE Dispenser
Color Clear
Dimensions (L x W x H) 4 x 18 x 20 in.
Holds PPE, Bouffant Caps, Isolation Gowns or Coveralls, Shoe Covers, Bouffant Caps
Material PETG
Mounting Wall Mount
No. of Compartments 3
Product Style Side Loading
Edges Rounded
For Use With (Application) PPE Isolation Station
For Use With (Equipment) PPE
Includes Mounting Hardware
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.