Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPFIA4 Monoclonal Antibody (3B1)
GREENER_CHOICE

Catalog No. PIMA524428
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524428 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524428 Supplier Invitrogen™ Supplier No. MA524428
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: RQVMEREFNN LLALGTDRKL DDGDDKVFRR APSWRKRFRP REHHGRGGML SASAETLPAG FRVSTLGTLQ PPPAPPKKIM PEAHSHYLYG HMLSAF.

PPFIA4, or liprin-alpha-4, belongs to the liprin-alpha gene family. See liprin-alpha-1 (LIP1, or PPFIA1; MIM 611054) for background on liprins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPFIA4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3B1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PPFIA4
Gene Accession No. O75335
Gene Alias 1110008G13Rik; AI448359; AI852265; C81506; KIAA0897; Liprin-alpha4; Liprin-alpha-4; LOW QUALITY PROTEIN: liprin-alpha-4; PPFIA4; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-4; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 4; PTPRF interacting protein alpha 4; PTPRF-interacting protein alpha-4
Gene Symbols PPFIA4
Host Species Mouse
Immunogen PPFIA4 (NP_055868, 604 a.a. ~ 699 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8497
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.