Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPHLN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558583
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558583 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558583 Supplier Invitrogen™ Supplier No. PA558583
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKPPLLDRPG EGSYNRYYSH VDYRDYDEGR SFSHDRRSGP PHRGDESGYR WTRDDHSASR QPEYRDMRDG FRRKSFYSSH YARERSPYKR DNTFFRESPV GRKDSPHSR Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

The protein encoded by this gene is one of the several proteins that become sequentially incorporated into the cornified cell envelope during the terminal differentiation of keratinocyte at the outer layers of epidermis. This protein interacts with periplakin, which is known as a precursor of the cornified cell envelope. The cellular localization pattern and insolubility of this protein suggest that it may play a role in epithelial differentiation and contribute to epidermal integrity and barrier formation. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPHLN1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPHLN1
Gene Accession No. Q8NEY8
Gene Alias CDC7 expression repressor; CR; Gastric cancer antigen Ga50; HSPC206; HSPC232; MGC48786; periphilin 1; periphilin-1; periphilin-1-like protein; Pphln1
Gene Symbols PPHLN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPHLN1. Recombinant protein control fragment (Product #RP-97735).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51535
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.