Learn More
Invitrogen™ PPIAP22 Polyclonal Antibody

Description
PA1-9551 detects PPIA3L in human samples. PA1-9551 has been successfully used in Western blot procedures. By Western blot, PA1-9551 detects a 23 kDa protein representing PPIA3L. PA1-9551 immunogen is a purified recombinant PPIA3L.3 corresponding to the chromosome 21 gene (Human).
Specifications
Specifications
| Antigen | PPIAP22 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PPIAP22 |
| Gene Accession No. | 0 |
| Gene Alias | peptidylprolyl isomerase A (cyclophilin A) like |
| Gene Symbols | PPIAP22 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.