Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIL6 Monoclonal Antibody (OTI2G3), TrueMAB™, OriGene
SDP

Catalog No. p-200012885
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0081 30 μL
50-167-5444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0081 Supplier OriGene Supplier No. TA504840S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PPIL6 Monoclonal antibody specifically detects PPIL6 in Human samples. It is validated for Western Blot

Specifications

Antigen PPIL6
Applications Western Blot
Classification Monoclonal
Clone OTI2G3
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PPIL6
Gene Accession No. Q8IXY8
Gene Alias 2900084F20Rik; bA425D10.6; Cyclophilin-like protein PPIL6; dJ919F19.1; peptidyl-prolyl cis-trans isomerase-like 6; peptidyl-prolyl cis-trans isomerase-like variant a; peptidylprolyl isomerase (cyclophilin)-like 6; peptidylprolyl isomerase like 6; PPIase; PPIL6; Probable inactive peptidyl-prolyl cis-trans isomerase-like 6; radial spoke 12 homolog; rotamase PPIL6; RSPH12
Host Species Mouse
Immunogen Full length human recombinant protein of human PPIL6 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285755
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.