Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPM1F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112664
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112664 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112664 Supplier Invitrogen™ Supplier No. PA5112664
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPM1F Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

PPM1F is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase can interact with Rho guanine nucleotide exchange factors (PIX), and thus block the effects of p21-activated kinase 1 (PAK), a protein kinase mediating biological effects downstream of Rho GTPases. Calcium/calmodulin-dependent protein kinase II gamma (CAMK2G/CAMK-II) is found to be one of the substrates of this phosphatase. The overexpression of this phosphatase or CAMK2G has been shown to mediate caspase-dependent apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPM1F
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PPM1F
Gene Accession No. P49593, Q8CGA0
Gene Alias 1110021B16Rik; 4933427B07Rik; Ca(2+)/calmodulin-dependent protein kinase phosphatase; calcium/calmodulin-dependent protein kinase phosphatase; caM-kinase phosphatase; CAMKP; CaMKPase; FEM-2; hFem-2; KIAA0015; mKIAA0015; Partner of PIX 2; partner of PIX2; POPX2; PP2C phosphatase; Ppm1f; Protein fem-2 homolog; protein phosphatase 1F; protein phosphatase 1F (PP2C domain containing); protein phosphatase, Mg2+/Mn2+ dependent 1F; protein phosphatase, Mg2+/Mn2+ dependent, 1F
Gene Symbols PPM1F
Host Species Rabbit
Immunogen Recombinant Human Protein phosphatase 1F protein (1-76AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68606, 9647
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.