Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584232 Supplier Invitrogen™ Supplier No. PA584232
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PASLVILNEH NPPEIDDKRG PNTQGELQNA SPKQRKQSVY TPPTI Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 84%.

Protein phosphatase-1.[supplied by OMIM, Feb 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R1C
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R1C
Gene Accession No. Q8WVI7
Gene Alias 4930565M23Rik; 4930579P15Rik; inhibitor-5 of protein phosphatase 1; IPP5; PP1 inhibitor protein 5; PPP1R1C; protein phosphatase 1 regulatory inhibitor subunit 1C; protein phosphatase 1 regulatory subunit 1A; protein phosphatase 1 regulatory subunit 1C; protein phosphatase 1, regulatory (inhibitor) subunit 1C; RGD1562095
Gene Symbols PPP1R1C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R1C. Recombinant protein control fragment (Product #RP-100782).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151242
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.