Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R2P3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104041 Supplier Invitrogen™ Supplier No. PA5104041
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PPP1R2P3.

Inhibitor of protein-phosphatase 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R2P3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPP1R2B
Gene Accession No. Q6NXS1
Gene Alias PPP1R2P3; protein phosphatase 1 regulatory inhibitor subunit 2 pseudogene 3
Gene Symbols PPP1R2B
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPP1R2P3(Accession Q6NXS1), corresponding to amino acid residues G62-P112.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153743
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.