Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R9A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501732229 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2229 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2229 Supplier Proteintech Group Inc Supplier No. 188011AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Neurabin I is a brain-specific protein which contains an F-actin binding domain, a PDZ domain, a transmembrane-protein-interacting-domain, and a coiled-coil region. As its structure suggests, neurabin I is a promiscuous protein binding to F-actin, protein phosphatase 1, TGN38, and p70 S6 kinase. Found exclusively in brain, this protein is highly concentrated in the synapse and has also been shown to be enriched in the lamellipodia of the growth cone during neuronal development. Neurabin I appears to be a bridging protein by targeting other proteins to the synapse or by linking membrane proteins to the actin cytoskeleton.

Specifications

Antigen PPP1R9A
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PPP1R9A
Gene Accession No. O35867, Q9ULJ8
Gene Alias KIAA1222, Neurabin 1, Neurabin I, NRB1, NRBI, PPP1R9A
Gene Symbols PPP1R9A
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243725, 55607, 84685
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.