Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R2A Monoclonal Antibody (F.722.1)
GREENER_CHOICE

Catalog No. PIMA515007 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515007 Supplier Invitrogen™ Supplier No. MA515007
Add to Cart
Edge
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the B-prime (PR61), B-prime-prime or B-prime-prime-prime families of PP2A B subunits.

The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an alpha isoform of the regulatory subunit B55 subfamily. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R2A
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone F.722.1
Concentration 42 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
Gene PPP2R2A
Gene Accession No. P36876, P63151, Q6P1F6
Gene Alias 2410004D02Rik; B55A; B55ALPHA; hypothetical protein; mKIAA1541; PP2A subunit B isoform alpha; PP2A subunit B isoform B55-alpha; PP2A subunit B isoform BRA; PP2A subunit B isoform PR55-alpha; PP2A subunit B isoform R2-alpha; PP2A, subunit B, B55-alpha isoform; PP2A, subunit B, B-alpha isoform; PP2A, subunit B, BRA isoform; PP2A, subunit B, PR55-alpha isoform; PP2A, subunit B, R2-alpha isoform; PPP2R2A; PR52A; PR55A; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform; protein phosphatase 2 regulatory subunit B, alpha; protein phosphatase 2 regulatory subunit Balpha; protein phosphatase 2, regulatory subunit B, alpha; protein phosphatase 2A 55 kDa regulatory subunit, alpha isoform; serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, alpha isoform; serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform; serine-threonine protein phosphatase 2A regulatory subunit; testicular tissue protein Li 156
Gene Symbols PPP2R2A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the sequence of human PP2A B subunit.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117104, 5520, 71978
Target Species Drosophila, Human, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.