Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R5C Monoclonal Antibody (TQ11-1G6)
GREENER_CHOICE

Catalog No. 393600
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-360-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-360-0 Supplier Invitrogen™ Supplier No. 393600
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

PPP2R5C Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Phosphatases are involved in the regulation of MAP kinases.Protein phosphatase 2A (PP2A) is a critical component of the pathway responsible for p38 MAPK up-regulation of SERT catalytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R5C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone TQ11-1G6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PPP2R5C
Gene Accession No. Q13362
Gene Alias 2610043M05Rik; 2700063L20Rik; AI060890; AW545884; B' alpha regulatory subunit; B55-GAMMA; B56/PP2A gamma; B56G; Band 8A; C85228; D12Bwg0916e; gamma isoform of regulatory subunit B56, protein phosphatase 2A; IMYPNO; IMYPNO1; KIAA0044; MGC23064; MGC33570; mKIAA0044; PP2A B subunit isoform B56-gamma; PP2A B subunit isoform B'-alpha-3; PP2A B subunit isoform B'-gamma; PP2A B subunit isoform PR61-gamma; PP2A B subunit isoform R5-gamma; Ppp2r5c; PR52; PR55G; PR61G; protein phosphatase 2 regulatory subunit B', gamma; protein phosphatase 2 regulatory subunit B'gamma; protein phosphatase 2, regulatory subunit B (B56), gamma isoform; protein phosphatase 2, regulatory subunit B', gamma; protein phosphatase 2, regulatory subunit B', gamma isoform; renal carcinoma antigen NY-REN-29; Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit gamma isoform
Gene Symbols PPP2R5C
Host Species Mouse
Immunogen specific to Human PP2A B56.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5527
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.