Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP3CB Mouse anti-Human, Mouse, Porcine, Rat, Clone: 3C1A3, Proteintech
SDP

Catalog No. 501737032 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7032 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7032 Supplier Proteintech Group Inc Supplier No. 666141IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PPP3CB is a calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin.

Specifications

Antigen PPP3CB
Applications Western Blot
Classification Monoclonal
Clone 3C1A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PPP3CB
Gene Accession No. P16298, P20651, P48453
Gene Alias CALNA2, CALNB, CAM PRP catalytic subunit, CNA2, PPP3CB
Gene Symbols Ppp3cb
Host Species Mouse
Immunogen PPP3CB Fusion Protein Ag26987
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19056, 24675, 396603, 5532
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.