Learn More
Description
PPP6C Polyclonal Antibody for Western Blot, IP
Specifications
Specifications
| Antigen | PPP6C |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | Ppp6c |
| Gene Accession No. | O00743, Q64620, Q9CQR6 |
| Gene Alias | 2310003C10Rik; ANKRD33; FLJ34236; PP6; PP6C; PPP6; PPP6C; Ppv; PP-V; protein phosphatase 6 catalytic subunit; protein phosphatase 6, catalytic subunit; protein phosphatase V; serine/threonine protein phosphatase catalytic subunit; serine/threonine-protein phosphatase 6 catalytic subunit; Serine/threonine-protein phosphatase 6 catalytic subunit, N-terminally processed; UNQ5837 |
| Gene Symbols | Ppp6c |
| Host Species | Sheep |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
