Learn More
Sartorius PQ-Q AND PQ-K BUNDLE
Supplier:  Sartorius 185140
Encompass
Performance qualification kits for Quantification Kinetics or Both Applications for Octet RH16 system. Includes 1 kit with all necessary reagents biosensors microplates manuals and USB stick for performing two instrument qualifications. Requires Octet BLI Discovery and Octet BLI analysis studio Software version 12.0 or higher.
Catalog No. 50-289-1761
Add to Cart
Request a Quote