Learn More
Clontech Labs 3P pQCXIX Retroviral Vector | 20 ug
SDP
Supplier:  Clontech Labs 3P 631515
Encompass
The pQCXIX Retroviral Vector is a self-inactivating retroviral expression vector that is optimized to eliminate promoter interference from the upstream LTR in the integrated provirus. It expresses two target genes from a single, bicistronic message. The CMV promoter present in the 5' LTR produces high viral titers in standard packaging cell lines. A deletion in the 3' LTR provides the self-inactivating feature after reverse transcription of the expression cassette. This vector is useful for applications in which high titers and high-level bicistronic gene expression are desired.
Catalog No. NC1544750
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.