Learn More
Sartorius PQK KIT FOR RED384
Supplier:  Sartorius 181161
Encompass
Performance qualification kits for Quantification Kinetics or Both Applications for Octet RH16 system. Includes 1 kit with all necessary reagents biosensors microplates manuals and USB stick for performing two instrument qualifications. Requires Octet BLI Discovery and Octet BLI analysis studio Software version 12.0 or higher.
Catalog No. 50-289-1712