Learn More
Abcam PR 619, DUB Inhibitor, >99% Purity, Solid, 25 mg
Description
- Molecular Weight: 223.3 Da
- Purity: >99%
- Form: Solid
- Inhibitor Type: Cell-permeable, non-selective, and reversible DUB inhibitor
- IC50: 5 to 20 μM
- Biological Activity: Induces polyubiquitinated protein accumulation in cells without directly affecting proteasome activity; activates autophagy
- Chemical Name: [2,6-diamino-5-(cyanosulfanyl)pyridin-3-yl]sulfanylformonitrile
- CAS Number: 2645-32-1
- Storage Temperature: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Solid |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Deubiquitinase (DUB) |
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Formulation | 100% {[2,6-diamino-5-(cyanosulfanyl)pyridin-3-yl]sulfanyl}formonitrile |
| Quantity | 25 mg |
| For Use With (Application) | DUB inhibition, autophagy activation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.