Learn More
Teledyne ISCO, Inc Pr Class Dual Piston Pump With Belt Drive And Shaped Cams; 0.00 - 300.00 Ml/Min. ; 1,000 Psi
Supplier:  Teledyne ISCO, Inc PR300KRX
Encompass
PR Class Dual Piston Pump with Belt Drive and Shaped Cams; 0.00 - 300.00 mL/min. ; 1,000 psi; Hastelloy Steel Fluid Path; Jacketed Head; Automatic Piston Wash; with Pulse Dampener, Pressure & Leak Sensors; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/4"""" tubing
Catalog No. 50-211-9959
Only null left
Add to Cart
Request a Quote