Learn More
Biocare Medical, LLC Progesterone Receptor (PR) [SP2] Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC ACA302A
Encompass_Preferred
The presence of progesterone receptor (PR) in breast tumors indicates an increased likelihood of response to anti-estrogen (tamoxifen) therapy. The Progesterone Receptor [SP2] antibody clone is a high affinity rabbit monoclonal. A study has shown that the SP2 clone had a much higher affinity as compared to mouse monoclonals for the progesterone receptor. Studies have also shown that the SP2 clone provides supplementary evidence to ER in predicting survival in human breast cancer.
Catalog No. 50-827-90
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.