Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PR3 Monoclonal Antibody (PR3G-2), FITC
GREENER_CHOICE

Catalog No. FERMA549093 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549093 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549093 Supplier Invitrogen™ Supplier No. MA549093
  • $785.00
    $660.65 / Each of 1

    Save $124.35

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The monoclonal antibody PR3G-2 was produced by immunization of mice with a crude granule extract.

Serine protease that degrades elastin, fibronectin, laminin, vitronectin, and collagen types I, III, and IV (in vitro) (PubMed:3198760, PubMed:2033050, PubMed:28240246). By cleaving and activating receptor F2RL1/PAR-2, enhances endothelial cell barrier function and thus vascular integrity during neutrophil transendothelial migration (PubMed:23202369). May play a role in neutrophil transendothelial migration, probably when associated with CD177 (PubMed:22266279). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PR3
Applications Flow Cytometry, Functional Assay, Inhibition Assays, Western Blot
Classification Monoclonal
Clone PR3G-2
Concentration 100 μg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.02% sodium azide
Gene PRTN3
Gene Accession No. P24158
Gene Alias ACPA; AGP7; azurophil granule protein 7; CANCA; C-ANCA; C-ANCA antigen; leukocyte proteinase 3; MBN; MBT; mPR3; Myeloblastin; neutrophil proteinase 4; NP4; NP-4; P29; PR3; PR-3; Proteinase 3; PRTN3; serine proteinase, neutrophil; wegener autoantigen; Wegener granulomatosis autoantigen
Gene Symbols PRTN3
Host Species Mouse
Immunogen Monoclonal antibody PR3G-2 reacts with human proteinase 3 (PR3), a 30 kDa protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5657
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.