Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Pramipexole dihydrochloride monohydrate, 98%

Catalog No. AC463220050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463220050 5 g
AC463220250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463220050 Supplier Thermo Scientific Chemicals Supplier No. 463220050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191217-81-9
Molecular Formula C10H21Cl2N3OS
Molecular Weight (g/mol) 302.26
MDL Number MFCD00876894,MFCD02183927
InChI Key APVQOOKHDZVJEX-QTPLPEIMSA-N
IUPAC Name dihydrogen (6S)-N6-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine hydrate dichloride
SMILES [H+].[H+].O.[Cl-].[Cl-].CCCN[C@H]1CCC2=C(C1)SC(N)=N2

Specifications

Assay Percent Range 97.5 % min. (HPLC)
Quantity 5 g
Solubility Information Solubility aq. soln.: soluble. Other solubilities: soluble in methanol, slightly soluble in ethanol, insoluble in dichloromethane
Formula Weight 302.27
Chemical Name or Material Pramipexole dihydrochloride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.