Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRAS40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120978
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120978 Supplier Invitrogen™ Supplier No. PA5120978
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse heart, Rat heart. The target is usually found in the following locations: Cytoplasm, cytosol. Immunogen sequence: STDDGSLSEE TPAGPPTCSV PPASALPTQQ YAKSLPVSVP VWGFKEKRTE ARSSDEENGP PSSPDLDRIA ASMRALVLRE AEDTQVFGDL PRPRLNTSDF QKLKRKY.

The Akt signaling pathway contributes to the regulation of apoptosis after a variety of cell death signals. AKT1S1, also known as PRAS40, is a proline-rich substrate of the kinase AKT1 and is thought to play a role in neuroprotection mediated by nerve growth factor (NGF) after transient focal cerebral ischemia. AKT1S1 is also a substrate and potential regulator of mammalian target of rapamycin (mTOR), a serine/threonine kinase that regulates cell growth and cell cycle, and a negative regulator of autophagy. Treatment with the insulin-like growth factor-1 (IGF1) can induce the phosphorylation of AKT1S1 via the PI3K/AKT signaling pathway in PC12 cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRAS40
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AKT1S1
Gene Accession No. Q96B36, Q9D1F4
Gene Alias 1110012J22Rik; 40 kDa proline-rich AKT substrate; AI227026; AKT1 substrate 1; AKT1 substrate 1 (proline rich); AKT1 substrate 1 (proline-rich); akt1s1; Lobe; Lobel; OTTHUMP00000196756; OTTHUMP00000196760; PKB/Akt substrate 40; Pras; PRAS40; Proline-rich AKT substrate; proline-rich AKT1 substrate 1
Gene Symbols AKT1S1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 150-256 of human AKT1S1/PRAS40 (NP_001092103.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292887, 67605, 84335
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.