Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Praseodymium(III) oxide, 99.9% (metals basis)

Catalog No. AA3566318
Click to view available options
Quantity:
10 g
50 g

CAS: 12036-32-7 | O3Pr2 | 329.812 g/mol

Praseodymium(III) oxide is used as a pigment in glass, optic and ceramic industries. Praseodymium-doped glass called didymium glass is used in welding goggles, since it blocks infra-red radiation. Further, it is used as a dielectric in combination with silicon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-32-7
Molecular Formula O3Pr2
Molecular Weight (g/mol) 329.812
MDL Number MFCD00011178
InChI Key MMKQUGHLEMYQSG-UHFFFAOYSA-N
Synonym dipraseodymium trioxide, praseodymium iii oxide, praseodymium oxide pr2o3, praseodynium oxide, dipraseodymium 3+ ion trioxidandiide, praseodymium iii oxide trace metals basis 10g
PubChem CID 165911
IUPAC Name oxygen(2-);praseodymium(3+)
SMILES [O-2].[O-2].[O-2].[Pr+3].[Pr+3]

Specifications

Melting Point 2300°C
Physical Form Sintered Lump
Quantity 50 g
Merck Index 14,7712
Solubility Information Insoluble in water. Moderately soluble in strong mineral acids.
Formula Weight 329.83
Percent Purity 99.9%
Odor Odorless
Density 7.07 g/mL
Assay (metals basis)
Chemical Name or Material Praseodymium(III) oxide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 234-857-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.