Learn More
Apexbio Technology LLC Prasugrel hydrochloride 389574-19-0 10mM (in 1mL DMSO)
Supplier: Apexbio Technology LLC B12835.1
Prasugrel hydrochloride (CAS 389574-19-0) is a small molecule inhibitor that targets platelet activation by antagonizing the P2Y12 adenosine diphosphate (ADP) receptor on platelet surfaces By inhibiting this receptor prasugrel impedes ADP-mediated platelet aggregation a critical process in thrombus formation In biochemical assays prasugrel hydrochloride demonstrates an IC50 of 1 8 M for platelet inhibition This compound is utilized in biomedical research to investigate platelet signaling pathways and to model antithrombotic interventions in vitro
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.