Learn More
Apexbio Technology LLC Ro 31-8220 125314-64-9 50mg
Supplier: Apexbio Technology LLC B128750
Ro 31-8220 (CAS 125314-64-9) is a potent and selective inhibitor of protein kinase C (PKC) targeting isoforms PKC- (IC50 5 nM) PKC- I (24 nM) PKC- II (14 nM) PKC- (27 nM) and PKC- (24 nM) PKC is a Ca2 and phospholipid-dependent serine/threonine kinase involved in signaling pathways regulating cell proliferation and malignancy Ro 31-8220 modulates downstream signaling by inducing c-Jun expression and activating JNK1 while inhibiting MKP-1 and c-Fos expression independent of PKC activity In immunological studies it suppresses IL-2 production (IC50 80 nM) T cell proliferation (IC50 350 nM) and CD25 expression indicating inhibition of both early and late T cell activation Ro 31-8220 is widely utilized in cellular and molecular studies to dissect PKC-dependent and independent signaling mechanisms
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.